Learn More
Biolegend FITC anti-human HLA-A2
Supplier:  Biolegend 343303
Encompass_Preferred
FITC anti-human HLA-A2 [BB7.2]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-168-174