Learn More
Biolegend FITC anti-human CD326 (EpCAM) Recombinant [QA20A75]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Supplier:  Biolegend 377505
Encompass_Preferred
FITC anti-human CD326 (EpCAM) Recombinant [QA20A75]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-3208