Learn More
Biolegend FITC anti-mouse CD183 CXCR3
Supplier:  Biolegend 126535
Encompass_Preferred
FITC anti-mouse CD183 (CXCR3) [CXCR3-173]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-272-2712