Learn More
Biolegend FITC anti-mouse CD117 (c-kit)
Supplier:  Biolegend 135115
Encompass_Preferred
FITC anti-mouse CD117 (c-kit) [ACK2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-207-1081