Learn More
Biolegend FITC anti-mouse CD19
Supplier:  Biolegend 115506
Encompass_Preferred
FITC anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-163-543