Learn More
Biolegend FITC anti-mouse CD279 (PD-1)
Supplier:  Biolegend 135214
Encompass_Preferred
FITC anti-mouse CD279 (PD-1) [29F.1A12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-164-827