Learn More
Biolegend FITC anti-mouse CD3
Supplier:  Biolegend 100203
Encompass_Preferred
FITC anti-mouse CD3 [17A2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-162-368