Learn More
Biolegend FITC anti-mouse CD3ε
Supplier:  Biolegend 100305
Encompass_Preferred
FITC anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-162-386