Learn More
Biolegend FITC anti-mouse CD45RB
Supplier:  Biolegend 103306
Encompass_Preferred
FITC anti-mouse CD45RB [C363-16A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-162-819