Learn More
Biolegend FITC anti-mouse I-Ad
Supplier:  Biolegend 115005
Encompass_Preferred
FITC anti-mouse I-Ad [39-10-8]; Isotype: Mouse (C3H.SW) IgG3, κ; Reactivity: Mouse d and g7 haplotypes; Apps: FC; Size: 50 ug
Catalog No. 50-163-522