Learn More
CPC Scientific FITC-(beta-Ala)-Ile-Thr-Phe-X-Asp-Leu-Leu-X-Tyr-Tyr-Gly-Pro-NH2 (with special cyclization to get double bond, X= (S)-alpha-(2?-pentenyl)alanine) 1MG
SDP
Supplier:  CPC Scientific STAP002B
Encompass
This NYAD-2 peptide is NYAD-1 labeled with FITC (Ex/Em=493/517 nm). NYAD-1 is a cell-penetrating alphalpha-Helical peptide used as a potential HIV-1 inhibitor. The hydrocarbon-stapled structure enhances alpha helicity.ONE-LETTER SEQUENCE: FITC-(beta-A)-ITFXDLLXYYGP-NH2 (with special cyclization to get double bond, X= (S)-alpha-(2?-pentenyl)alanine)MOLECULAR FORMULA: MOLECULAR WEIGHT:1911.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: AntimicrobialREFERENCES: Zhang, H. et al. J. Mol. Biol. 378, 565 (2008)
Catalog No. 50-211-0446