Learn More
Medchemexpress LLC FITC-Chitosan | 95.0% | 100000 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYD2826100MG
Encompass_Preferred
FITC-Chitosan (MW 100000) is a Chitosan labeled with FITC. It can be used for fluorescent labeling and imaging.
  • Used for fluorescent labeling and imaging.
Catalog No. 50-004-29618