Learn More
Medchemexpress LLC FITC-Dextran | 60842-46-8 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY128868H100MG
Encompass_Preferred
FITC-Dextran (MW 500000) is a fluorescent dye utilized in biomedical research as a tracer molecule. It combines fluorescein isothiocyanate (FITC) with dextran, a carbohydrate, to form a stable fluorescent tracer. This allows for labeling and tracking cells or other biological matter, visible under microscopy or quantifiable with specialized detection instruments.
  • Acts as a fluorescent probe (Ex=495 nm; Em=525 nm)
  • Can reveal heat shock-induced cell damage
  • Supports studies on early and late stages of apoptosis
  • Suitable for cell permeability studies
  • Assesses blood-brain barrier disruption
Catalog No. 50-003-38205