Learn More
Apexbio Technology LLC FITC-Dextran (MW 150000) 10mg
SDP
Supplier:  Apexbio Technology LLC A906610
Encompass_Preferred
FITC-Dextran MW 150000 (CAS 60842-46-8) is a fluorescent polysaccharide probe composed of dextran covalently labeled with fluorescein isothiocyanate (FITC) It is designed as a macromolecular marker to study cell membrane permeability and barrier integrity in biological systems FITC-Dextran MW 150000 functions by emitting detectable green fluorescence after excitation enabling quantitative analysis of transport dynamics and permeability Based on these properties FITC-Dextran MW 150000 is employed in research applications such as permeability assays evaluation of barrier integrity (blood-brain intestinal vascular) intracellular trafficking studies and assessment of fluid-phase uptake processes
Catalog No. 50-000-95917