Learn More
Medchemexpress LLC HY-128868A 5mg Medchemexpress, FITC-Dextran (MW 4000) CAS:60842-46-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128868A5MG
Encompass_Preferred
Medchemexpress, HY-128868A 5mg FITC-Dextran (MW 4000) CAS:60842-46-8 FITC-Dextran (MW 4000) is a marker consisting of coupling fluorescein-isothiocyanate to dextran (4 kDa). FITC-Dextran are polysaccharides comprised of varying lengths of branched glucose molecules and can be used to determine solute, ion and protein permeability of the blood-brain barrier (BBB) based on the size of the dextran used[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0140