Learn More
ACROBiosystems FITC-Labeled Human Syndecan-1 Protein, His Tag
Supplier:  ACROBiosystems SD1HF228200UG
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human Syndecan-1, His Tag (SD1-HF228) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Gly 254 (Accession # P18827-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 42-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Syndecan-1 antibody, Mouse IgG1 at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human Syndecan-1, His Tag (Cat. No. SD1-HF228) with a linear range of 0.078-2.5 μg/mL (QC tested).
Catalog No. 50-205-5183