Learn More
ABclonal Technology FITC Rabbit anti-Human CD7 mAb
SDP
Supplier:  ABclonal Technology A26834200T
Encompass_Preferred
This gene encodes a transmembrane protein which is a member of the immunoglobulin superfamily This protein is found on thymocytes and mature T cells It plays an essential role in T-cell interactions and also in T-cell B-cell interaction during early lymphoid development
Catalog No. 50-000-11489