Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) APC22004 Quick-disconnect insert, acetal, 1/4"" tubing ID, pack of 10
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0636092
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-583