Learn More
Waters Corp Fitting, Reducing Union, 1/4 in. FNPT, 1/8 in. V
Supplier:  Waters Corp 700006646
Encompass_Preferred
This fitting is a reducing union designed to connect 1/8 in. stainless steel tubing to a CO2 supply tank, commonly found in systems like the Bio-Botanical Extraction System. It features a 1/4 in. FNPT connection on one end and a 1/8 in. V connection on the other, facilitating a secure and efficient union.
  • Serves as a reducing union.
  • Connects 1/8 in. stainless steel tubing.
  • Suitable for CO2 supply tanks.
  • Features 1/4 in. FNPT x 1/8 in. V connections.
  • Used in systems such as the Bio-Botanical Extraction System.
Catalog No. 50-239-1280