Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Straight, Hose Barb Union, 5/16"" ID; 100/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3062588
Encompass
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. 50-629-236