Learn More
Abcam Fi x & Perm / Cell Fi x ation & Permeabilization Kit (Flow Cytometry), 200 Tests
Supplier:  Abcam AB185917200TEST
Encompass_Preferred
Fix & Perm / Cell fixation and permeabilization kit is designed to improve the staining of intracellular markers for flow cytometry by fixing cells and permeabilizing cell membranes, enabling access to cytoplasmic, mitochondrial, nuclear, and other organelle markers. - Optimizes antibody access to intracellular structures while maintaining cell morphology - Reduces background while allowing simultaneous addition of permeabilization medium and fluorochrome-labeled antibodies - Quick and easy: incubate with fixation medium, followed by permeabilization medium and antibody, with minimal washing required
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0279