Learn More
Enzo Life Sciences Fixation Permeabilization, (buffer) (100 ml)
SDP
Supplier:  Enzo Life Sciences ADI950010100
Encompass_Preferred
Formulation: Liquid. PBS, pH 7.2, containing 4% formaldehyde and 0.1% saponin. Applications: Flow Cytometry. Shipping: Blue Ice Not Frozen. Long Term Storage: +4°C
Catalog No. 50-658-671