Learn More
Med Vet International Fixation Pins End, Positive Thread, J0371F, Small, Each
Supplier:  Med Vet International JOR371F
Encompass_Preferred
Small. Shaft 3/32" dia., Thread 1/8" dia., Thread Pitch 1"L, Total 4"L, Thread Pitch 0.9mm
Catalog No. 50-005-81519