Learn More
Ethos Biosciences Fixative Histology pathology selective 1/EA
Supplier:  Ethos Biosciences 334832
Encompass_Preferred
B-5 Fixative is a mercury-based fixative formulated for pathology and hematology applications requiring excellent nuclear and cytoplasmic detail Commonly used for bone marrow lymph node and hematopoietic tissue specimens it provides superior preservation for diagnostic evaluation and special staining procedures The solution delivers consistent fixation for reliable morphology and clear microscopic results B-5 Fixative is suitable for routine and specialized laboratory workflows where high-quality tissue detail is essential Supplied in chemically compatible containers it is appropriate for clinical research and reference laboratory environments
Catalog No. 50-005-74315