Learn More
Ethos Biosciences Zinc Sulfate, 1gal, Ethos
Supplier:  Ethos Biosciences 6845G
Encompass_Preferred
Zinc Sulfate is a chemical reagent suitable for microbiology, parasitology, and general laboratory applications. Commonly used in flotation techniques for the concentration and recovery of parasitic organisms, it supports reliable specimen preparation and microscopic evaluation. The formulation provides consistent composition for accurate and reproducible results. It is also suitable for reagent preparation, chemical analysis, and other routine laboratory procedures. Supplied in chemically compatible containers, Zinc Sulfate is appropriate for clinical, research, and educational laboratory environments.
Catalog No. 50-005-74130