Learn More
Trajan Scientific FIXED NEEDLE SYRINGE
SDP
Supplier:  Trajan Scientific 0019821
Encompass_Preferred
Trajan Fixed-Needle Autosampler Syringes for Thermo Scientific Instruments, Diameter Outer Needle: 0.47 mm, 0.018 in., Length Needle: 50 mm, 1.96 in., Model: 5F-BT-5/0.47C, Needle Gauge: 26 G, Volume: 5 uL, Connection Type: Fixed
Catalog No. 50-253-0410