Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Value Collection Fixed Sewer Nozzle, 4200 psi, 6mm Orifice

Supplier:  MSC 59814749

Encompass

Value Collection Fixed Sewer Pressure Washer Nozzle is designed for efficient cleaning in sewer applications. It features a robust design for reliable performance.

  • Sewer nozzle type
  • Fixed nozzle style
  • Withstands maximum pressure of 4200 psi
  • Features a 1/4 in thread size
  • Equipped with a 6mm orifice diameter
  • Manufactured in Switzerland

Catalog No. 50-283-6299


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.