Learn More
Perkin Elmer US LLC Fixed-Angle Specular Reflectance Accessory
Supplier:  Perkin Elmer US LLC 01860796
Encompass_Preferred
Good quality spectra can be obtained from films on glossy flat surfaces, shiny paper, metallic surfaces, and semiconductor wafers such as silicon and germanium using the PerkinElmer Fixed-Angle Specular Reflectance Accessory. You can also obtain high-quality spectra of surface-treated materials, coatings, thinly lubricated surfaces, resin, and molded plastic surfaces. In semiconductor technology, Fixed-Angle Specular Reflectance is the technique of choice used to map the thickness of epitaxial layers on semiconductor wafers. Benefits Include: Easy to mount in sample compartment Three interchangeable aperture masks provided for studying small sample areas Optimized optical design and easy installation
Catalog No. 50-211-7253