Learn More
Cayman Chemical FK-506-13C-d2 500ug
SDP
Supplier:  Cayman Chemical 22178500 UG
Encompass_Preferred
An internal standard for the quantification of FK-506 by GC- or LC-MS
Catalog No. 50-298-5698