Learn More
Medchemexpress LLC FKB04 | 99.98% | 363.20 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY169120
Encompass_Preferred
FKB04 is a telomeric repeat binding factor 2 (TRF2) inhibitor that disrupts the telomere maintenance mechanism in liver cancer cells. This leads to T-loop defects, telomere shortening, and cellular senescence, thereby exerting antitumor activity. It can also inhibit tumor growth in a human liver cancer xenograft mouse model. This compound is intended for liver cancer research.
  • Disrupts telomere maintenance mechanism in liver cancer cells
  • Leads to T-loop defects, telomere shortening, and cellular senescence
  • Exerts antitumor activity
  • Inhibits tumor growth in a human liver cancer xenograft mouse model
  • Used in liver cancer research
Catalog No. 50-003-91880