Learn More
Medchemexpress LLC Fkbp12 protac rc32 | 2375555-66-9 | 97.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13083550MG
Encompass_Preferred
FKBP12 PROTAC RC32 is a potent FKBP12 degrader based on PROTAC technology. This degrader combines Rapamycin and a Cereblon E3 ubiquitin ligase ligand, Pomalidomide. It achieves approximately 50% protein degradation (DC50) of FKBP12 at 0.3 nM in Jurkat cells within 12 hours. In vivo, it effectively degrades FKBP12 in most organs of treated mice, Bama pigs, and rhesus monkeys, with oral administration also showing significant degradation in mice.
  • Potent FKBP12 degrader based on PROTAC technology
  • Composed of Rapamycin and a Cereblon E3 ubiquitin ligase ligand
  • Achieves 50% protein degradation (DC50) of approximately 0.3 nM in Jurkat cells within 12 hours
  • Degrades FKBP12 protein in most organs of treated mice (except brain)
  • Efficiently degrades FKBP12 in various organs of Bama pigs and rhesus monkeys
  • Effective with oral administration in mice
Catalog No. 50-003-40324