Learn More
Enzo Life Sciences FKBP59 monoclonal antibody (KN382/EC1) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISRA1400D
Encompass_Preferred
Clone: KN382/EC1. Host: Mouse. Isotype: IgG1. Immunogen: Partially purified 9S rabbit uterine progesterone receptor. Source: Purified from mouse ascites. Species reactivity: Human, Mouse Rabbit, Monkey, Mink, Hamster, Guinea pig, Bovine. Applications: IHC, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7671