Learn More
Medchemexpress LLC Fl118-14-propanol | 2821768-98-1 | 450.44 g/mol | C24H22N2O7 | 5 MG

Supplier: Medchemexpress LLC HY1568565MG
FL118-14-Propanol is a camptothecin-derived compound used as an antibody-drug conjugate (ADC) payload in preclinical cancer research. An ADC synthesized with this compound has shown antitumor activity in mice. It is supplied as a solid and is available as a DMSO solution for research applications.
- Used as an ADC payload for preclinical antitumor studies.
- Molecular formula: C24H22N2O7.
- Molecular weight: 450.44 g/mol.
- Available as solids in multiple sizes and as a 10 mM solution in DMSO.
- For research use only; not validated for clinical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.