Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fl118-14-propanol | 2821768-98-1 | 98.7% | 450.44 | C24H22N2O7 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY156856100MG

Encompass_Preferred

FL118-14-Propanol is a small-molecule derivative of FL118 used in preclinical research as an ADC payload with reported anti-tumor activity in mouse models. It is supplied as a solid or solution for research use only.

  • Cas number: 2821768-98-1.
  • Molecular formula: C24H22N2O7.
  • Molecular weight: 450.44 g/mol.
  • High listed purity: 98.7%.
  • Available formats: solid (multiple mg sizes) and 10 mM solutions in dmsO.
  • Intended use: research only, commonly used as an ADC cytotoxin payload in preclinical studies.

Catalog No. 50-002-04481


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.