Learn More
Medchemexpress LLC Fl118-C3-O-C-amide-C-nh2 formate | 95.8% | 582.56 g/mol | C28H30N4O10 | 10 MG

Supplier: Medchemexpress LLC HY161022A10MG
FL118-C3-O-C-amide-C-NH2 formate is an ADC linker supplied as a solid formate salt for research use in the synthesis of antibody-drug conjugates. It is hygroscopic, highly soluble in DMSO with ultrasonic assistance, and requires low-temperature, moisture-controlled storage to maintain stability.
- Appearance: solid, white to yellow.
- Purity: 95.82%.
- Molecular weight: 582.56 g/mol.
- Chemical formula: C28H30N4O10.
- Solubility: soluble in DMSO at 100 mg/mL with ultrasonic assistance.
- Storage: store sealed at -20°C (powder); in solvent keep at -80°C up to 6 months.
- Intended use: for research use only; not for clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.