Learn More
ATCC FL74-UCD-1 (CRL-8012) Feline Lymphoblastoid Cell Line, Suspension Culture
Supplier: ATCC CRL8012
FL74-UCD-1 is a feline lymphoblastoid cancer cell line established from a cat with lymphoma. The line grows in suspension, exhibits lymphoblast morphology, and is persistently infected with a feline leukemia virus strain that produces high viral titers with attenuated virulence. It is used in virology, vaccine development, and immunology research.
- Organism: Felis catus (cat).
- Cell type: Lymphoblastoid cancer cell line.
- Morphology: Lymphoblast; suspension culture.
- Disease origin: Feline lymphoma.
- Viral status: Persistently infected with feline leukemia virus; produces high viral titers with attenuated virulence.
- Typical applications: Virology, vaccine production, immunology.
- Accession/RRID: CVCL_6391.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.