Learn More
Medchemexpress LLC Fladrafinil | 90212-80-9 | 98.6% | 25 MG

Supplier: Medchemexpress LLC 25MGHY129048
Fladrafinil (CRL-40,941) is a nootropic compound primarily intended for research use only. It is not sold for patient use.
- Functions as a nootropic
- Suitable for research applications
- Powder stable for 3 years at -20°C
- Powder stable for 2 years at 4°C
- Solution stable for 6 months at -80°C
- Solution stable for 1 month at -20°C
- Appears as a white to off-white solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.