Learn More
Cederlane FLAER: Alexa Fluor 488 proaerolysin FL
SDP
Supplier:  Cederlane FL2
Encompass
FLAER: Alexa Fluor 488 proaerolysin FL2 contains 50 μg of lyophilized FLAER. When reconstituted in 0.5 ml or 1.0 ml of PBS-0.05% Sodium Azide respectively a 10 -6 M FLAER stock solution is produced. These solutions also contain 5 mg/mL of albumin, added in order to maximize stability. This item is labeled for clinical use only
Catalog No. NC1637041