Learn More
CAPRICO BIOTECHNOLOGIES INC Flaer iFluor 488 RUO
Supplier:  CAPRICO BIOTECHNOLOGIES INC 3105114
Encompass_Preferred
The Flaer iFluor 488 RUO is a fluorescent dye used for labeling proteins and other biomolecules for research applications. It has an excitation maximum of 494 nm and emission maximum of 518 nm. The dye is optimized for minimal self-quenching and high brightness. It is compatible with common filters and has good photostability. The dye is supplied as a lyophilized solid that is soluble in water and buffers. It can be conjugated to antibodies, proteins, peptides, and other biomolecules using NHS-ester or maleimide chemistry. The dye is suitable for applications like flow cytometry, fluorescence microscopy, cell labeling, and protein labeling.
Catalog No. NC2194625