Learn More
CPC Scientific H-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MISC006A
Encompass
Peptide that is comprised of of eight amino acids (DYKDDDDK), representing a versatile fusion tag with regards to recombinant proteins purification. This epitope tag is a short hydrophilic, highly charged peptide that is the most widely used epitope tag employed in structural and functional studies of proteins. It is commonly used for eluting immunoaffinity chromatography-purified recombinant proteins.SEQUENCE: H-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: DYKDDDDKMOLECULAR FORMULA: C41H60N10O20MOLECULAR WEIGHT: 1013STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [98849-88-8]SYNONYMS: FLAG Epitope PeptideRESEARCH AREA: MiscellaneousCATEGORY: Miscellaneous Peptides CITATIONS:Lee, Jung-Rok, et. al. ACS Nano 10, no. 12 (2016):
Catalog No. 50-210-9729