Learn More
TARGETMOL CHEMICALS INC FLAG PEPTIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TP127325MG
Encompass_Preferred
Also available in 5 mg 10 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FLAG peptide is an eight amino acids peptide (Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys) with an enterokinase-cleavage site used for the purification of recombinant proteins. purity: 100%
Catalog No. 50-257-8862