Learn More
Lakeland Industries Flame Resistant Coveralls
Supplier:  Lakeland Industries C065DH137030
Encompass_Preferred
Lakeland's flame resistant 6.5 oz Westex DH coverall provides you with inherent FR protection, advanced moisture management, and superior comfort. Dual certified to NFPA 2112 and NFPA 70E (CAT 2), this coverall is ideal for when protection against arch flash and flash fire is required.
Catalog No. 50-284-0517