Learn More
Chemglass Life Sciences Flange, Compatible with Buchi Rotary Evaporators, Flange OD: 145mm, Flange ID: 119mm, Shank OD: 130mm
SDP
Supplier:  Chemglass Life Sciences CG14610
Encompass_Preferred
Flange, Compatible with Buchi Rotary Evaporators, Flange OD: 145mm, Flange ID: 119mm, Shank OD: 130mm. Replacement flange compatible with Buchi condensers and rotary evaporator flasks.
Catalog No. 50-153-1114