Learn More
IDEX Health & Science Upchurch Super Flangeless Nut 6-40 Flat-Bottom, 1/16" OD, Green, 10 Pack
Supplier:  IDEX Health & Science Upchurch M64403X
Encompass_Preferred
Headless Super Flangeless Nut eliminate loosening of fittings due to tube twisting, holds tight even during vibration and delivers the highest pressure holding for flat bottom fittings that we offer. It is for a 1/16OD tubing and a 6-40 Flat-Bottom port.. This is a 10 pack.
Catalog No. NC3236523