Learn More
Microsolv Technology Corporation Flanging Tool Kit with 5 Drills, Tubing Cutter, Tubing Connectors, Tubing & Plastic Carrying Case. 1 EA.
Supplier:  Microsolv Technology Corporation 45099002
Encompass_Preferred
"Flangy the Tool" for cold flanging of Polymer or thin walled PEEK Tubing, provides an easy and smooth way to make or restore, tubing tip, flanges easy. As the flange is created by mechanical means rather than by heat, the resulting flange itself will be more long lasting and stable. This Kit includes the Flanging Tool, set of 5 flanging drills (pins), a sample of PTFE Tubing that is 1/16" ID x 0.75" OD, six feet long, and the Clean Cut Tubing Cutting Tool, a selection Tubing Connectors (nuts) with PP rings and the carrying case.
Catalog No. 50-227-1417