Learn More
Perkin Elmer US LLC Flared Solva MP2 Two-Stop Peristaltic Pump Tubing - 0.76 mm I.D., Black/Black, Pkg. 12
Supplier:  Perkin Elmer US LLC N8145216
Encompass_Preferred
This MP2 Two-Stop Peristaltic Pump Tubing is designed for use with the MP2 pump on your ICP-OES and ICP-MS instrument. The MP2 is a compact, precise, stand-alone micro peristaltic pump with a flow rate range from 0.20 µL/min to 10 mL/min and is available with one to eight channels. Optimized for low-flow rates, the MP2 uses either MP2 tubing or standard 3-stop pump tubing.
Catalog No. 50-210-5079