Learn More
Perkin Elmer US LLC Flared Solvent Resistant Two-Stop Peristaltic Pump Tubing - 0.76 mm I.D., Black/Black, Pkg. 12
Supplier:  Perkin Elmer US LLC N0811914
Encompass_Preferred
This flared end tubing to allow ease of connection to metal T, greatly reducing the time to replace peripump tubing.
Catalog No. 50-211-3313