Learn More
LUKNOVA INC Empty Column, 57 Ml Volume To Fit 40 Gram Silica, Luer Inlet, Polypropylene Structure, Polyethylene Frit Pore Size 20 µm, Chamber Ø 30 X 170 Mm. 20 Per Pack.
Supplier:  LUKNOVA INC FC0030401
Encompass
Luknova Patented Supersep Empty Flash Column, 57 Ml Volume Able To Fit 40 Gram Of Standard Silica (230-400 Mesh), Luerlock Inlet, Polypropylene Structure, Polyethylene Frit With Pore Size 20 µm, Empty Chamber Size Ø 30 X 170 Mm. These Columns Are Disposable And Each Unit Comes At 20 Columns Per Pack . They Are Designed To Be Assembled Easily And Fit Directly In The Teledyne Isco, Biotage Isolera, Interchim Puriflash, Yamazen, Santai, Agela, And Other Flash Systems. Great To Pack Your Own Columns With Youe Own Or Special Packing Materials. More Top And Bottom Frits Are Available.
Catalog No. 50-918-469