Learn More
Medchemexpress LLC FlAsH-EDT2 | 212118-77-9 | MFCD27977924 | 99.0% | C24H18As2O5S4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13041210MG
Encompass_Preferred
FlAsH-EDT2 is a fluorescein-based biarsenical probe that selectively binds tetracysteine (CCXXCC) motifs in proteins. It is membrane-permeant and largely nonfluorescent until it binds the target sequence, whereupon it produces strong green fluorescence suitable for live-cell imaging and in vitro biochemical assays. The compound contains arsenical groups; follow manufacturer safety and handling guidance.
Catalog No. 50-002-60792