Learn More
Deltatrak Inc FlashLink Sgl Use BLE Data Logger
SDP
Supplier:  Deltatrak Inc 40909
Encompass_Preferred
Sold by the case, 10 units per case. The FlashLink In-Transit BLE Loggers is a PDF logger with enhanced BLE capabilities and communications. The logger is ideal for wirelessly monitoring temperature of perishables as they travel through the cold chain, from produce to prepared foods, to chemicals and pharmaceuticals. Data is downloaded from the BLE Logger using a smart phone app. The mobile device uploads information to a web application where it can be viewed remotely.Receivers quickly and easily access the trip history using the mobile app without having to open cargo doors. Trip data is made available in graph and table format, and when the mobile app is synchronized to the web app, location is marked at the time the logger is interrogated.A PDF report is automatically generated in the app and can be easily shared via e-mail with other stakeholders. As a backup feature, the logger can also be read using its built-in USB port which automatically generates a PDF, when connected.
Catalog No. NC1880928